Assay Details
| Gene Symbol: Stambp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024239.2 | NP_077201.1 | 5 - 6 | 856 | 53 | |
| GenBank mRNA | AB010123.1 | - | 5 - 6 | 803 | 53 | |
| AK017600.1 | - | 5 - 6 | 811 | 53 | ||
| AK019907.1 | - | 5 - 6 | 806 | 53 | ||
| AK046995.1 | - | 5 - 6 | 807 | 53 | ||
| AK136961.1 | - | 5 - 6 | 1407 | 53 | ||
| AK139391.1 | - | 5 - 6 | 831 | 53 | ||
| AK168810.1 | - | 5 - 6 | 829 | 53 | ||
| AK220541.1 | - | 3 - 4 | 7087 | 53 | ||
| BC003497.1 | - | 5 - 6 | 799 | 3153829 | 53 | |
| BC006939.1 | - | 5 - 6 | 854 | 2649718 | 53 | |
| BC025111.1 | - | 5 - 6 | 798 | 4194332 | 53 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Stambp
Genomic Map