Assay Details
Gene Symbol: Khdrbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011317.4 | NP_035447.3 | 7 - 8 | 1313 | 85 | |
NR_045036.1 | 7 - 8 | 1313 | 85 | |||
GenBank mRNA | AK044927.1 | - | 7 - 8 | 1310 | 85 | |
AK050520.1 | - | 7 - 8 | 1308 | 85 | ||
AK146815.1 | - | 7 - 8 | 1275 | 85 | ||
AK150687.1 | - | 7 - 8 | 1292 | 85 | ||
AK152084.1 | - | 7 - 8 | 1292 | 85 | ||
AK204780.1 | - | 2 - 3 | 236 | 85 | ||
BC002051.1 | - | 7 - 8 | 1269 | 3490667 | 85 | |
U17046.1 | - | 7 - 8 | 1270 | 85 | ||
U17961.1 | - | 7 - 8 | 1281 | 85 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
