Assay Details
| Gene Symbol: Arhgap18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176837.2 | NP_789807.1 | 11 - 12 | 1643 | 137 | |
| GenBank mRNA | AK078522.1 | - | 6 - 7 | 695 | 137 | |
| AK162202.1 | - | 7 - 8 | 886 | 137 | ||
| BC030858.1 | - | 11 - 12 | 1643 | 4194996 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap18
Genomic Map