Assay Details
| Gene Symbol: Ppp1r32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133689.1 | NP_598450.1 | 7 - 8 | 787 | 70 | |
| GenBank mRNA | AK005821.1 | - | 7 - 8 | 820 | 70 | |
| AK016325.1 | - | 7 - 8 | 815 | 70 | ||
| AY032666.1 | - | 7 - 8 | 787 | 70 | ||
| BC118523.1 | - | 7 - 8 | 763 | 40105249 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r32
Genomic Map