Assay Details
| Gene Symbol: Gpsm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029522.2 | NP_083798.2 | 8 - 9 | 1459 | 60 | |
| GenBank mRNA | AK040996.1 | - | 8 - 9 | 1348 | 60 | |
| AK143552.1 | - | 8 - 9 | 1460 | 60 | ||
| AY081187.1 | - | 8 - 9 | 1303 | 60 | ||
| BC021308.1 | - | 7 - 8 | 1014 | 5007832 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpsm2
Genomic Map