Assay Details
Gene Symbol: Pxmp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008993.2 | NP_033019.2 | 2 - 3 | 388 | 101 | |
GenBank mRNA | AF309644.1 | - | 2 - 3 | 242 | 101 | |
AK003118.1 | - | 2 - 3 | 242 | 101 | ||
BC057975.1 | - | 2 - 3 | 229 | 5055267 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
