Assay Details
Gene Symbol: Prkg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008926.4 | NP_032952.3 | 13 - 14 | 1803 | 82 | |
GenBank mRNA | AK038739.1 | - | 13 - 14 | 1787 | 82 | |
AK081422.1 | - | 13 - 14 | 1804 | 82 | ||
BC113205.1 | - | 13 - 14 | 1714 | 40047751 | 82 | |
L12460.1 | - | 13 - 14 | 2075 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
