Assay Details
Gene Symbol: Rpl34-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199350.1 | NP_001186279.1 | 1 | 87 | 95 | |
GenBank mRNA | BC082285.1 | - | 1 | 82 | 95 | |
BC100360.1 | - | 1 | 87 | 30847080 | 95 |
Gene Symbol: Rpl34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005859.3 | NP_001005859.1 | 2 - 3 | 156 | 95 | |
NM_001287581.1 | NP_001274510.1 | 2 - 3 | 224 | 95 | ||
NM_026724.2 | NP_081000.1 | 2 - 3 | 292 | 95 | ||
GenBank mRNA | AK003177.1 | - | 2 - 3 | 93 | 95 | |
AK146252.1 | - | 2 - 3 | 100 | 95 | ||
BC028517.1 | - | 1 - 2 | 1496108 | 95 | ||
BC058118.1 | - | 2 - 3 | 263 | 5683990 | 95 | |
CJ041925.1 | - | 2 - 3 | 100 | 95 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
