Assay Details
| Gene Symbol: Cox15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144874.4 | NP_659123.2 | 2 - 3 | 367 | 66 | |
| GenBank mRNA | AK048099.1 | - | 2 - 3 | 349 | 66 | |
| AK051792.1 | - | 2 - 3 | 265 | 66 | ||
| AK141631.1 | - | 2 - 3 | 299 | 66 | ||
| BC011509.1 | - | 2 - 3 | 327 | 3981950 | 66 | |
| BC021498.1 | - | 2 - 3 | 287 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cox15
Genomic Map