Assay Details
| Gene Symbol: Por | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC003240.1 | - | 4 - 5 | 469 | 3497749 | 73 |
| BC031463.1 | - | 4 - 5 | 414 | 3497979 | 73 | |
| D17571.1 | - | 4 - 5 | 413 | 73 | ||
More Information
Set Membership: |
|

Por
Genomic Map