Assay Details
| Gene Symbol: Pla2g1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011107.1 | NP_035237.1 | 2 - 3 | 220 | 67 | |
| GenBank mRNA | AF094610.1 | - | 2 - 3 | 213 | 67 | |
| AF097637.1 | - | 2 - 3 | 201 | 67 | ||
| AF162712.1 | - | 2 - 3 | 220 | 67 | ||
| AF187852.1 | - | 2 - 3 | 199 | 67 | ||
| AK007730.1 | - | 2 - 3 | 214 | 67 | ||
| AK007797.1 | - | 2 - 3 | 219 | 67 | ||
| AK008331.1 | - | 2 - 3 | 216 | 67 | ||
| AK008664.1 | - | 2 - 3 | 216 | 67 | ||
| AK008668.1 | - | 2 - 3 | 216 | 67 | ||
| AK008841.1 | - | 2 - 3 | 214 | 67 | ||
| AK008934.1 | - | 2 - 3 | 220 | 67 | ||
| AK008936.1 | - | 2 - 3 | 214 | 67 | ||
| AK028104.1 | - | 2 - 3 | 217 | 67 | ||
| AK028134.1 | - | 2 - 3 | 220 | 67 | ||
| BC145908.1 | - | 2 - 3 | 220 | 40131211 | 67 | |
| BC145910.1 | - | 2 - 3 | 220 | 40131213 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pla2g1b
Genomic Map