Assay Details
Gene Symbol: Gm19744 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK032387.1 | - | 3 - 4 | 3276 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | teneurin transmembrane protein 3 | Tenm3 |
2610100B16Rik, Odz1, Odz3, Ten-m3, mKIAA1455 |
Anti-sense | Chr. 8 - 48225665 - 48935162 on NCBI Build GRCm38 | |
Downstream | RIKEN cDNA 1700019L22 gene | 1700019L22Rik | 226.8kb | Anti-sense | Chr. 8 - 49096147 - 50918746 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 461630 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610079 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510695 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 137289 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 384727 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 109894 | - | - |
Whole Mouse Genome G4122A | A_52_P939208 | AK032387.1 | mouse_transcriptome |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850679 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850627 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850646 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850650 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850660 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06998791 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850664 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850657 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850642 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850761 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850661 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850683 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850643 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850734 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850776 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850750 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850703 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850774 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850720 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850799 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850645 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850671 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850751 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850781 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850757 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850770 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850782 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850663 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850689 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850699 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850727 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850665 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850670 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06691410 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850655 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850628 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850706 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850728 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850731 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850789 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850668 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850717 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06691408 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07064138 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850721 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850653 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850637 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07034292 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850771 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850756 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850639 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850696 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850708 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850644 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850697 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07034273 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850695 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850631 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850735 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850712 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850691 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850715 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850649 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850777 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850772 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850794 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850800 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850623 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850659 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850677 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850674 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850634 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850792 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850684 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850711 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850767 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850762 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850719 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850718 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850641 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850749 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850709 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07411298 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850753 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07080797 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850775 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850760 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850733 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850632 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850675 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850685 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850713 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850744 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850654 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850626 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850687 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850759 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850693 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06941427 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850796 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850778 | - | - |
