Assay Details
| Gene Symbol: Dram2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025582.2 | NP_001020753.1 | 5 - 6 | 602 | 83 | |
| GenBank mRNA | AK008532.1 | - | 5 - 6 | 526 | 83 | |
| BC024458.1 | - | 5 - 6 | 528 | 4975302 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dram2
Genomic Map