Assay Details
Gene Symbol: Rbm15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045807.1 | NP_001039272.1 | 1 - 2 | 3074 | 80 | |
GenBank mRNA | AK048987.1 | - | 2309 | 80 | ||
AK077048.1 | - | 1 - 2 | 3464 | 80 | ||
AK168242.1 | - | 1 - 2 | 2835 | 80 | ||
AK168936.1 | - | 1 - 2 | 2959 | 80 | ||
BC028452.1 | - | 1 - 2 | 632 | 80 | ||
BC051409.1 | - | 1 - 2 | 2828 | 80 | ||
BC057038.1 | - | 1 - 2 | 2838 | 80 | ||
BC080828.1 | - | 1 - 2 | 2819 | 80 | ||
BC120590.1 | - | 1 - 2 | 3074 | 40129513 | 80 | |
BC137741.1 | - | 1 - 2 | 3074 | 8860760 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
patterning of blood vessels
positive regulation of transcription of Notch receptor target
negative regulation of myeloid cell differentiation
spleen development
ventricular septum morphogenesis
placenta blood vessel development
protein binding
poly(A) RNA binding
negative regulation of transcription from RNA polymerase II promoter
patterning of blood vessels
positive regulation of transcription of Notch receptor target
negative regulation of myeloid cell differentiation
spleen development
ventricular septum morphogenesis
placenta blood vessel development
protein binding
poly(A) RNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405715 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405706 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405703 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405704 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405711 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405768 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405767 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405712 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405718 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405720 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405705 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405701 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405721 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405719 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405707 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405700 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405708 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405714 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405702 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405713 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405709 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405717 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P430262 | BC137741.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P430262 | BC120590.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P430262 | BC057038.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P430262 | BC080828.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P430262 | BC028452.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P430262 | BC137741.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P430262 | BC120590.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P430262 | BC057038.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P430262 | BC080828.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P430262 | BC028452.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 2101 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 947655 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 548075 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 637700 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 505005 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 737552 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 18432 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177266 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171677 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P430262 | BC137741.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P430262 | BC120590.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P430262 | BC057038.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P430262 | BC080828.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P430262 | BC028452.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P430262 | BC137741.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P430262 | BC120590.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P430262 | BC057038.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P430262 | BC080828.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P430262 | BC028452.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00721296 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177266 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171677 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177266 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171677 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405714 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405705 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405713 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405715 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405706 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405700 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405720 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405702 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405767 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405701 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405703 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405712 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405708 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405718 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405717 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405768 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405711 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405709 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405704 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405721 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405707 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22405719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405611 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405731 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405639 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405673 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405706 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405695 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405682 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405722 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405690 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405705 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405683 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405680 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405668 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405699 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405710 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405728 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405691 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405617 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405605 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405656 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405643 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405651 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405724 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405712 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405714 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405637 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405700 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405645 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405663 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405700 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405714 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02302041 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405716 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405741 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03076271 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405731 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405657 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405659 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405680 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405670 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405649 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02353336 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405687 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405738 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02353347 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405698 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405653 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405743 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405684 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405751 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405715 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405692 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03174076 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405732 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405740 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405708 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405705 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405677 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405702 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405710 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02353352 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405734 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405646 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405735 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405749 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405691 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405754 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02630862 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405647 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405750 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405753 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405661 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405656 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405742 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405728 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405712 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405752 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405737 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405695 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405678 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405724 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405651 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405699 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405682 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02403920 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405673 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405719 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405668 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405675 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405706 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405726 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02630856 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10501093 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10501094 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1434514_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10501091 | - | - |
Mouse Genome 430 2.0 Array | 1434514_at | - | - |
Mouse Genome 430 2.0 Array | 1434514_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 229700_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 229700_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0300021054.mm.1 | NM_001045807.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300021054.mm.1 | NM_001045807.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300021054.mm.1 | TC0300002706.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300021050.mm.1 | NM_001045807.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300021050.mm.1 | NM_001045807.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300021050.mm.1 | TC0300002706.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300021053.mm.1 | NM_001045807.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300021053.mm.1 | NM_001045807.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300021053.mm.1 | TC0300002706.mm.1 | MTA TC |
Mouse Expression Set 430 | 1434514_PM_at | - | - |
Mouse Expression Set 430 | 1434514_at | - | - |
Murine Genome U74v2 Set | 161002_PM_at | - | - |
Murine Genome U74v2 Set | 161002_at | - | - |
Mouse Genome 430A 2.0 Array | 1434514_at | - | - |
Mouse Genome 430A 2.0 Array | 1434514_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1434514_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1434514_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 850039 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 730286 | AB290927.1 | genbank_mrna_mouse |
MouseWG-6 v2 Expression BeadChip | 730286 | AF271381.1 | genbank_mrna_mouse |
MouseWG-6 v2 Expression BeadChip | 730286 | AK046772.1 | genbank_mrna_mouse |
MouseWG-6 v2 Expression BeadChip | 730286 | BC046386.1 | genbank_mrna_mouse |
MouseWG-6 v2 Expression BeadChip | 730286 | BC116297.2 | genbank_mrna_mouse |
MouseWG-6 v2 Expression BeadChip | 730286 | BC116323.2 | genbank_mrna_mouse |
MouseWG-6 v2 Expression BeadChip | 730286 | NM_181422.2 | refseq_mrna_mouse_NM |
MouseRef-8 v2 Expression BeadChip | ILMN_2945275 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2945275 | - | - |
