Assay Details
Gene Symbol: Pear1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032413.1 | NP_001027585.1 | 11 - 12 | 131 | ||
NM_001032414.1 | NP_001027586.1 | 10 - 11 | 131 | |||
NM_001289600.1 | NP_001276529.1 | 10 - 11 | 131 | |||
NM_001289601.1 | NP_001276530.1 | 12 - 13 | 131 | |||
NM_028460.2 | NP_082736.1 | 12 - 13 | 131 | |||
NM_152799.2 | NP_690012.2 | 12 - 13 | 131 | |||
GenBank mRNA | AF440279.1 | - | 12 - 13 | 131 | ||
AK053551.1 | - | 11 - 12 | 131 | |||
AK141082.1 | - | 131 | ||||
AK161049.1 | - | 12 - 13 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
