Assay Details
Gene Symbol: Bap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027088.2 | NP_081364.1 | 4 - 5 | 388 | 85 | |
GenBank mRNA | AB047820.1 | - | 4 - 5 | 289 | 85 | |
AK079374.1 | - | 4 - 5 | 404 | 85 | ||
AK129108.1 | - | 4 - 5 | 296 | 85 | ||
AK170576.1 | - | 4 - 5 | 389 | 85 | ||
AK171561.1 | - | 4 - 5 | 390 | 85 | ||
BC042489.1 | - | 4 - 5 | 339 | 85 | ||
BC050901.1 | - | 4 - 5 | 351 | 6402443 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
