Assay Details
Gene Symbol: Banf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038231.2 | NP_001033320.1 | 1 - 2 | 99 | 62 | |
GenBank mRNA | AK010257.1 | - | 1 - 2 | 85 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK010257.1, NM_001038231.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
