Assay Details
Gene Symbol: Prkd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171004.1 | NP_001164475.1 | 8 - 9 | 1934 | 67 | |
NM_001171005.1 | NP_001164476.1 | 7 - 8 | 1130 | 67 | ||
NM_029239.3 | NP_083515.2 | 8 - 9 | 1934 | 67 | ||
GenBank mRNA | AK030058.1 | - | 7 - 8 | 1127 | 67 | |
BC027320.1 | - | 7 - 8 | 1333 | 67 | ||
BC037012.1 | - | 7 - 8 | 1578 | 5342428 | 67 | |
BC089164.1 | - | 8 - 9 | 1833 | 5400250 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
