Assay Details
Gene Symbol: Parp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145619.2 | NP_663594.2 | 5 - 6 | 781 | 92 | |
GenBank mRNA | AF368233.1 | - | 5 - 6 | 781 | 92 | |
AK044223.1 | - | 5 - 6 | 774 | 92 | ||
AK088125.1 | - | 5 - 6 | 783 | 92 | ||
AK145259.1 | - | 5 - 6 | 815 | 92 | ||
AK147868.1 | - | 5 - 6 | 1080 | 92 | ||
AK165070.1 | - | 4 - 5 | 869 | 92 | ||
AK170541.1 | - | 5 - 6 | 725 | 92 | ||
AK209984.1 | - | 1 - 2 | 101 | 92 | ||
BC014870.1 | - | 5 - 6 | 721 | 3602116 | 92 | |
BC058754.1 | - | 5 - 6 | 701 | 6476689 | 92 | |
BC062204.1 | - | 5 - 6 | 701 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
