Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100022177.mm.1 |
NM_001252515.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022177.mm.1 |
NM_001252515.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022177.mm.1 |
NM_153078.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022177.mm.1 |
NM_153078.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022177.mm.1 |
TC1100002334.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100022172.mm.1 |
NM_001252515.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022172.mm.1 |
NM_001252515.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022172.mm.1 |
NM_153078.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022172.mm.1 |
NM_153078.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022172.mm.1 |
TC1100002334.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100022173.mm.1 |
NM_001252515.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022173.mm.1 |
NM_001252515.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022173.mm.1 |
NM_153078.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022173.mm.1 |
NM_153078.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022173.mm.1 |
TC1100002334.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100022174.mm.1 |
NM_001252515.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022174.mm.1 |
NM_001252515.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022174.mm.1 |
NM_153078.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022174.mm.1 |
NM_153078.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022174.mm.1 |
TC1100002334.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100022175.mm.1 |
NM_001252515.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022175.mm.1 |
NM_001252515.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022175.mm.1 |
NM_153078.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022175.mm.1 |
NM_153078.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022175.mm.1 |
TC1100002334.mm.1 |
MTA TC |