Assay Details
Gene Symbol: Acy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276442.1 | NP_001263371.1 | 2 - 3 | 208 | 58 | |
NM_025371.3 | NP_079647.1 | 2 - 3 | 212 | 58 | ||
GenBank mRNA | AK002423.1 | - | 2 - 3 | 198 | 58 | |
AK003703.1 | - | 2 - 3 | 213 | 58 | ||
BC005631.1 | - | 2 - 3 | 216 | 3964138 | 58 | |
CV560337.1 | - | 2 - 3 | 133 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
