Assay Details
Gene Symbol: Gpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173866.3 | NP_776291.1 | 5 - 6 | 665 | 95 | |
GenBank mRNA | AK031958.1 | - | 5 - 6 | 857 | 95 | |
AK033424.1 | - | 5 - 6 | 666 | 95 | ||
AK075894.1 | - | 5 - 6 | 684 | 95 | ||
AK076250.1 | - | 5 - 6 | 683 | 95 | ||
AK082030.1 | - | 5 - 6 | 708 | 95 | ||
BC034219.1 | - | 5 - 6 | 697 | 5065322 | 95 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
