Assay Details
Gene Symbol: Aurka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011497.4 | NP_035627.1 | 1 - 2 | 179 | 68 | |
GenBank mRNA | AF007817.1 | - | 1 - 2 | 126 | 68 | |
BC014711.1 | - | 1 - 2 | 115 | 3968835 | 68 | |
CD560646.1 | - | 1 - 2 | 102 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
