Assay Details
Gene Symbol: March10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039242.2 | NP_001034331.1 | 5 - 6 | 738 | 92 | |
GenBank mRNA | AK132734.1 | - | 5 - 6 | 727 | 92 | |
BC100474.1 | - | 5 - 6 | 731 | 30918723 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
