Assay Details
Gene Symbol: Dera | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172733.1 | NP_766321.1 | 5 - 6 | 544 | 105 | |
GenBank mRNA | AK012558.1 | - | 5 - 6 | 580 | 105 | |
AK034347.1 | - | 546 | 105 | |||
AK075963.1 | - | 5 - 6 | 569 | 105 | ||
AK083298.1 | - | 5 - 6 | 544 | 105 | ||
AK145820.1 | - | 5 - 6 | 580 | 105 | ||
AK150273.1 | - | 5 - 6 | 570 | 105 | ||
AK150712.1 | - | 5 - 6 | 540 | 105 | ||
AK152462.1 | - | 5 - 6 | 548 | 105 | ||
AK168204.1 | - | 5 - 6 | 580 | 105 | ||
AK169418.1 | - | 5 - 6 | 564 | 105 | ||
BC016218.1 | - | 5 - 6 | 550 | 4484163 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
