Assay Details
| Gene Symbol: Hdac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010411.2 | NP_034541.2 | 5 - 6 | 495 | 87 | |
| GenBank mRNA | AF074881.1 | - | 5 - 6 | 442 | 87 | |
| AK145158.1 | - | 5 - 6 | 461 | 87 | ||
| BC139300.1 | - | 5 - 6 | 523 | 8862322 | 87 | |
| BC139301.1 | - | 5 - 6 | 523 | 8862323 | 87 | |
| BQ932476.1 | - | 5 - 6 | 507 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hdac3
Genomic Map