Assay Details
Gene Symbol: Faf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007983.2 | NP_032009.2 | 3 - 4 | 556 | 83 | |
GenBank mRNA | AK183710.1 | - | 2 - 3 | 121 | 83 | |
AK186332.1 | - | 2 - 3 | 121 | 83 | ||
BC065098.1 | - | 3 - 4 | 556 | 30362116 | 83 | |
U39643.1 | - | 3 - 4 | 566 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
