Assay Details
Gene Symbol: Gnl3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168600.1 | NP_001162071.1 | 5 - 6 | 420 | 108 | |
NM_198110.2 | NP_932778.1 | 5 - 6 | 453 | 108 | ||
GenBank mRNA | BB644039.1 | - | 5 - 6 | 453 | 108 | |
BC020354.1 | - | 5 - 6 | 389 | 108 | ||
BC057033.1 | - | 5 - 6 | 379 | 6834283 | 108 | |
BC079653.1 | - | 5 - 6 | 375 | 6843880 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
