Assay Details
Gene Symbol: Polr1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009085.2 | NP_033111.2 | 7 - 8 | 893 | 67 | |
GenBank mRNA | AK132114.1 | - | 7 - 8 | 835 | 67 | |
AK146240.1 | - | 7 - 8 | 843 | 67 | ||
BC130018.1 | - | 7 - 8 | 894 | 40140532 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
