Assay Details
Gene Symbol: Gfm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146043.2 | NP_001139515.1 | 16 - 17 | 1688 | 110 | |
NM_001271463.1 | NP_001258392.1 | 16 - 17 | 1688 | 110 | ||
NM_001271464.1 | NP_001258393.1 | 15 - 16 | 1607 | 110 | ||
NM_001271465.1 | NP_001258394.1 | 16 - 17 | 1715 | 110 | ||
NM_177266.5 | NP_796240.3 | 16 - 17 | 1721 | 110 | ||
GenBank mRNA | AK035819.1 | - | 16 - 17 | 1626 | 110 | |
AK044371.1 | - | 15 - 16 | 1608 | 110 | ||
AK083441.1 | - | 15 - 16 | 1726 | 110 | ||
AK161797.1 | - | 16 - 17 | 1624 | 110 | ||
AK165871.1 | - | 16 - 17 | 1682 | 110 | ||
BC027341.1 | - | 16 - 17 | 1672 | 3984129 | 110 | |
BC057062.1 | - | 5 - 6 | 1499 | 110 | ||
BC138233.1 | - | 16 - 17 | 1619 | 8861252 | 110 | |
BC145208.1 | - | 16 - 17 | 1619 | 9053759 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
