Assay Details
Gene Symbol: Srgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081011.2 | NP_001074480.2 | 8 - 9 | 1777 | 84 | |
GenBank mRNA | BC030457.1 | - | 7 - 8 | 1084 | 84 | |
BC151081.1 | - | 7 - 8 | 1395 | 9087994 | 84 | |
BC151082.1 | - | 7 - 8 | 1395 | 9087995 | 84 | |
BC158055.1 | - | 7 - 8 | 1393 | 9088163 | 84 | |
BC172152.1 | - | 7 - 8 | 1395 | 9054822 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
