Assay Details
| Gene Symbol: Rgsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243223.1 | NP_001230152.1 | 16 - 17 | 2829 | 73 | |
| GenBank mRNA | AK029630.1 | - | 7 - 8 | 936 | 73 | |
| AK161463.1 | - | 9 - 10 | 1249 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2023593 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 193450 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 597732 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 453696 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 803463 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913653 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 413 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105425 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167143 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P112664 | AK029630.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2023593 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00220282 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04312484 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P167143 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105425 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04312466 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105425 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P167143 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728722 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728850 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728768 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00402686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728842 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00435095 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728710 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728765 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728808 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00435094 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728743 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00291499 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00613400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728812 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728763 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00567807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728826 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728715 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00433733 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728779 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00549372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00402679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00282266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728814 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00606183 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00402683 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728741 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728830 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00613416 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00402670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728731 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728759 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00282264 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00613415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728747 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728724 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00893610 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00115435 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01113203 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728754 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728778 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728758 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728711 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00688527 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728744 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00982745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00079723 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728819 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00291500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00606175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01113202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728817 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00606188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728772 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728766 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728793 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00537181 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728728 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00402676 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728706 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728822 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728816 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00435089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00613409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00533484 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00115434 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728865 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10358910 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10358911 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10358912 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100026710.mm.1 | NM_001243223.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100026710.mm.1 | NM_001243223.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100026710.mm.1 | TC0100003282.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100026712.mm.1 | NM_001243223.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100026712.mm.1 | NM_001243223.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100026712.mm.1 | TC0100003282.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100026711.mm.1 | NM_001243223.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100026711.mm.1 | NM_001243223.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100026711.mm.1 | TC0100003282.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100026717.mm.1 | NM_001243223.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100026717.mm.1 | NM_001243223.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100026717.mm.1 | TC0100003282.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100026713.mm.1 | NM_001243223.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100026713.mm.1 | NM_001243223.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100026713.mm.1 | TC0100003282.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100026709.mm.1 | NM_001243223.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100026709.mm.1 | NM_001243223.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100026709.mm.1 | TC0100003282.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 434298 | - | - |
| ABI Mouse Genome Survey Microarray | 774339 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1220565 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1220565 | - | - |

Rgsl1
Genomic Map