Assay Details
Gene Symbol: Asb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023046.4 | NP_075533.3 | 1 - 2 | 93 | 69 | |
GenBank mRNA | AK006515.1 | - | 1 - 2 | 94 | 69 | |
BC049699.1 | - | 1 - 2 | 93 | 69 | ||
BU938650.1 | - | 1 - 2 | 96 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK006515.1, NM_023046.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
