Assay Details
Gene Symbol: 4933407L21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_037692.1 | 1 - 2 | 607 | 144 | ||
GenBank mRNA | AK016730.1 | - | 1 - 2 | 606 | 144 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | integral membrane protein 2C | Itm2c |
3110038L02Rik, BRI3, Bricd2c, E25, E25C, ITM3 |
-19.8kb | Sense | Chr. 1 - 85894510 - 85908698 on NCBI Build GRCm38 |
Downstream | G protein-coupled receptor 55 | Gpr55 |
Gm218, Lpir1 |
2.7kb | Anti-sense | Chr. 1 - 85933206 - 85991567 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P5905 | AK037440.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P5905 | AW125058.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P5905 | NR_037692.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P5905 | AK016730.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P5905 | AK037440.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P5905 | AW125058.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P5905 | NR_037692.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P5905 | AK016730.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100131 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P5905 | AK037440.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P5905 | AW125058.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P5905 | NR_037692.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P5905 | AK016730.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P5905 | AK037440.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P5905 | AW125058.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P5905 | NR_037692.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P5905 | AK016730.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100131 | - | - |
Whole Mouse Genome G4122A | A_52_P5905 | AK037440.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P5905 | AW125058.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P5905 | NR_037692.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P5905 | AK016730.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P5905 | AK037440.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P5905 | AW125058.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P5905 | NR_037692.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P5905 | AK016730.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P325889 | AK037440.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100131 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20409217 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00088515 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00088496 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20409253 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00088514 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20409222 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00088481 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00088510 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00088508 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00088509 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20409267 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00501039 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00088495 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00644853 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01016681 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20409262 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00501048 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20409213 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00088488 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00088501 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20409229 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20409255 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00501037 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20409246 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00088486 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20409258 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20409256 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00088482 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20409252 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00088506 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20409225 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20409202 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01079766 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00501041 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1430206_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1430206_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1430206_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71141_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71141_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0100006460.mm.1 | NR_037692.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100006460.mm.1 | NR_037692.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100006460.mm.1 | TC0100000775.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100006458.mm.1 | NR_037692.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100006458.mm.1 | NR_037692.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100006458.mm.1 | TC0100000775.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100006462.mm.1 | NR_037692.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100006462.mm.1 | NR_037692.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100006462.mm.1 | TC0100000775.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100006459.mm.1 | NR_037692.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100006459.mm.1 | NR_037692.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100006459.mm.1 | TC0100000775.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100006465.mm.1 | NR_037692.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100006465.mm.1 | NR_037692.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100006465.mm.1 | TC0100000775.mm.1 | MTA TC |
Mouse Expression Set 430 | 1430206_at | - | - |
Mouse Expression Set 430 | 1430206_PM_at | - | - |
Murine Genome U74v2 Set | 129895_at | - | - |
Murine Genome U74v2 Set | 129895_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1430206_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1430206_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 733034 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2742223 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2742223 | - | - |
