Assay Details
Gene Symbol: Cnot6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290741.1 | NP_001277670.1 | 2 - 3 | 300 | 84 | |
NM_212484.2 | NP_997649.1 | 3 - 4 | 669 | 84 | ||
GenBank mRNA | AK181033.1 | - | 1 - 2 | 30 | 84 | |
AK184045.1 | - | 1 - 2 | 30 | 84 | ||
AK217278.1 | - | 1 - 2 | 30 | 84 | ||
AY043269.1 | - | 2 - 3 | 298 | 84 | ||
BC062950.1 | - | 3 - 4 | 657 | 6840162 | 84 | |
CF746424.1 | - | 1 - 2 | 134 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
