Assay Details
Gene Symbol: Ddx59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026500.3 | NP_080776.1 | 3 - 4 | 1126 | 140 | |
GenBank mRNA | AK004833.1 | - | 3 - 4 | 1127 | 140 | |
AK133731.1 | - | 3 - 4 | 1088 | 140 | ||
BC023840.1 | - | 3 - 4 | 1053 | 5344890 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
