Assay Details
Gene Symbol: Lgals9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159301.1 | NP_001152773.1 | 2 - 3 | 233 | 115 | |
NM_010708.2 | NP_034838.2 | 2 - 3 | 233 | 115 | ||
GenBank mRNA | AK146044.1 | - | 2 - 3 | 189 | 115 | |
BC003754.1 | - | 2 - 3 | 160 | 3601419 | 115 | |
CA458877.1 | - | 2 - 3 | 223 | 115 | ||
CA546274.1 | - | 2 - 3 | 233 | 115 | ||
U55060.1 | - | 2 - 3 | 159 | 115 | ||
U55061.1 | - | 2 - 3 | 140 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
