Assay Details
Gene Symbol: Nras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010937.2 | NP_035067.2 | 5 - 6 | 748 | 73 | |
GenBank mRNA | AK010412.1 | - | 5 - 6 | 709 | 73 | |
AK011704.1 | - | 5 - 6 | 695 | 73 | ||
AK150357.1 | - | 5 - 6 | 687 | 73 | ||
AK165960.1 | - | 5 - 6 | 692 | 73 | ||
BC014704.1 | - | 5 - 6 | 676 | 73 | ||
BC051443.1 | - | 5 - 6 | 671 | 30061371 | 73 | |
BC058755.1 | - | 5 - 6 | 680 | 6475312 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
