Assay Details
Gene Symbol: Endou | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168693.1 | NP_001162164.1 | 4 - 5 | 521 | 110 | |
NM_008902.3 | NP_032928.1 | 3 - 4 | 520 | 110 | ||
GenBank mRNA | AK132616.1 | - | 4 - 5 | 462 | 110 | |
AK169457.1 | - | 2 - 3 | 611 | 110 | ||
M95545.1 | - | 3 - 4 | 520 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
