Assay Details
Gene Symbol: Rcbtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170694.1 | NP_001164165.1 | 10 - 11 | 1204 | 66 | |
NM_134083.4 | NP_598844.3 | 10 - 11 | 1199 | 66 | ||
NR_033185.1 | 11 - 12 | 1584 | 66 | |||
GenBank mRNA | AK013540.1 | - | 1309 | 66 | ||
AK031242.1 | - | 10 - 11 | 1165 | 66 | ||
AK031248.1 | - | 10 - 11 | 1170 | 66 | ||
AK147784.1 | - | 11 - 12 | 1205 | 66 | ||
AK160881.1 | - | 11 - 12 | 1391 | 66 | ||
BC003224.1 | - | 8 - 9 | 997 | 3482729 | 66 | |
JN086338.1 | - | 9 - 10 | 1120 | 66 | ||
JN086339.1 | - | 10 - 11 | 1358 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
