Assay Details
Gene Symbol: Rcbtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170694.1 | NP_001164165.1 | 6 - 7 | 635 | 68 | |
NM_134083.4 | NP_598844.3 | 6 - 7 | 630 | 68 | ||
NR_033185.1 | 7 - 8 | 1015 | 68 | |||
GenBank mRNA | AK013540.1 | - | 738 | 68 | ||
AK031242.1 | - | 6 - 7 | 596 | 68 | ||
AK031248.1 | - | 6 - 7 | 601 | 68 | ||
AK147784.1 | - | 7 - 8 | 636 | 68 | ||
AK160881.1 | - | 7 - 8 | 822 | 68 | ||
BC003224.1 | - | 4 - 5 | 428 | 3482729 | 68 | |
JN086338.1 | - | 5 - 6 | 551 | 68 | ||
JN086339.1 | - | 6 - 7 | 789 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
