Assay Details
Gene Symbol: Mark1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145515.2 | NP_663490.2 | 16 - 17 | 2065 | 84 | |
GenBank mRNA | AB185282.1 | - | 16 - 17 | 1991 | 84 | |
AK173181.1 | - | 15 - 16 | 1918 | 84 | ||
BC112400.1 | - | 16 - 17 | 1994 | 40059725 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
