Assay Details
Gene Symbol: Slc9b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178877.6 | NP_849208.4 | 2 - 3 | 218 | 93 | |
GenBank mRNA | AK076505.1 | - | 3 - 4 | 258 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
