Assay Details
| Gene Symbol: Setdb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081024.1 | NP_001074493.1 | 13 - 14 | 2534 | 127 | |
| GenBank mRNA | BC157926.1 | - | 12 - 13 | 2356 | 9088099 | 127 |
| BC172021.1 | - | 12 - 13 | 2356 | 9054307 | 127 | |
| EU155090.1 | - | 12 - 13 | 2348 | 127 | ||
| EU155092.1 | - | 12 - 13 | 2366 | 127 | ||
| EU155093.1 | - | 11 - 12 | 2282 | 127 | ||
| EU155094.1 | - | 11 - 12 | 2282 | 127 | ||
| EU155096.1 | - | 11 - 12 | 2005 | 127 | ||
| EU155097.1 | - | 13 - 14 | 2407 | 127 | ||
| EU155098.1 | - | 11 - 12 | 2300 | 127 | ||
| EU155099.1 | - | 12 - 13 | 2364 | 127 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Setdb2
Genomic Map