Assay Details
Gene Symbol: Gpatch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026367.4 | NP_080643.4 | 8 - 9 | 1386 | 65 | |
GenBank mRNA | AK029990.1 | - | 8 - 9 | 1370 | 65 | |
AK030026.1 | - | 8 - 9 | 1385 | 65 | ||
AK083471.1 | - | 7 - 8 | 543 | 65 | ||
AK169660.1 | - | 9 - 10 | 1507 | 65 | ||
BC054810.1 | - | 8 - 9 | 1358 | 6839419 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
