Assay Details
Gene Symbol: 4930425O10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040545.1 | 1 - 2 | 126 | 108 | ||
GenBank mRNA | AK015203.1 | - | 1 - 2 | 126 | 108 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | eukaryotic translation initiation factor 4E | Eif4e |
EG668879, Eif4e-ps, If4e, eIF-4E |
-157.9kb | Anti-sense | Chr. 3 - 138526191 - 138557599 on NCBI Build GRCm38 |
Downstream | tetraspanin 5 | Tspan5 |
2810455A09Rik, 4930505M03Rik, NET-4, Tm4sf9 |
15.2kb | Anti-sense | Chr. 3 - 138742208 - 138904433 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
