Assay Details
Gene Symbol: Nup35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027091.4 | NP_081367.1 | 1 - 2 | 119 | 73 | |
GenBank mRNA | AK009187.1 | - | 1 - 2 | 67 | 73 | |
AK089997.1 | - | 1 - 2 | 118 | 73 | ||
AK136109.1 | - | 1 - 2 | 82 | 73 | ||
AK145775.1 | - | 1 - 2 | 66 | 73 | ||
BC048814.1 | - | 1 - 2 | 74 | 2647417 | 73 | |
BY272877.1 | - | 1 - 2 | 119 | 73 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
