Assay Details
Gene Symbol: Nr5a1os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK017050.1 | - | 1 - 2 | 139 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
adhesion G protein-coupled receptor D2, pseudogene |
Adgrd2-ps |
EG666416, Gm13585, Gm999, Gpr144, Gpr144-ps, OTTMUSG00000012858 |
-22.9kb | Sense | Chr. 2 - 38668953 - 38689748 on NCBI Build GRCm38 |
Downstream |
nuclear receptor subfamily 6, group A, member 1 |
Nr6a1 |
1700113M01Rik, Gcnf, NCNF, RTR |
2.7kb | Anti-sense | Chr. 2 - 38723369 - 38927688 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
