Assay Details
| Gene Symbol: Anapc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175300.4 | NP_780509.2 | 8 - 9 | 1708 | 104 | |
| GenBank mRNA | AK033784.1 | - | 8 - 9 | 1676 | 104 | |
| AK129351.1 | - | 8 - 9 | 1661 | 104 | ||
| AK154465.1 | - | 8 - 9 | 1662 | 104 | ||
| AK170676.1 | - | 8 - 9 | 1674 | 104 | ||
| AK196194.1 | - | 1 - 2 | 56 | 104 | ||
| BC027351.1 | - | 8 - 9 | 1656 | 104 | ||
| BC038267.1 | - | 7 - 8 | 1020 | 103 | ||
| BC096381.1 | - | 7 - 8 | 1901 | 104 | ||
| BC137583.1 | - | 8 - 9 | 1676 | 8860602 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Anapc2
Genomic Map