Assay Details
Gene Symbol: Unc13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029873.2 | NP_001025044.2 | 11 - 12 | 1532 | 74 | |
GenBank mRNA | AK147408.1 | - | 6 - 7 | 1045 | 74 | |
AY753536.1 | - | 11 - 12 | 1422 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
